N-(3-Dimethylaminopropyl)methacrylamide 95% stabilized

Supplier: Ambeed
Warning

Synonyms: N-(3-(Dimethylamino)propyl)methacrylamide

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A265132-25G A265132-500G
76722-904EA 6.37 USD
76722-904 76722-906
N-(3-Dimethylaminopropyl)methacrylamide 95% stabilized
N-(3-Dimethylaminopropyl)methacrylamide

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O
MW: 170.26 g/mol
MDL Number: MFCD00038359
CAS Number: 5205-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR