Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate 95%

Supplier: Ambeed
Warning

A263292-100MG A263292-1G A263292-250MG
76722-878EA 50 USD
76722-878 76722-880 76722-882
Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate 95%
Methyl (5-butyl-1H-benzimidazol-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₇N₃O₂
MDL Number: MFCD00864534
CAS Number: 14255-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR