Rasagiline mesylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-N-Propargyl-1-indanamine Methanesulphonate, (R)-N-(Prop-2-yn-1-yl)-2,3-dihydro-1H-inden-1-amine methanesulphonate, (R)-N-(2-Propyn-1-yl)-1-indanamine Methanesulphonate
A262921-100G
A262921-1G
A262921-1MG
A262921-250MG
A262921-50MG
A262921-5G
76722-854EA
1624
USD
76722-854
76722-856
76722-858
76722-860
76722-862
76722-864
Rasagiline mesylate 97%
Rasagiline mesylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₃N·CH₄O₃S |
MDL Number:
MFCD08460604 CAS Number: 161735-79-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...