Ethyl 4-hydroxy-6-methoxyquinoline-3-carboxylate 98%

Supplier: Ambeed
Warning

A262745-10G A262745-1G A262745-250MG A262745-25G A262745-5G
76722-844EA 65 USD
76722-844 76722-846 76722-848 76722-850 76722-852
Ethyl 4-hydroxy-6-methoxyquinoline-3-carboxylate 98%
Ethyl 4-hydroxy-6-methoxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO₄
MDL Number: MFCD00085617
CAS Number: 77156-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR