Stearamide 98%

Supplier: Ambeed
Warning

Synonyms: Octadecanamide, Stearic acid amide

A262686-100G A262686-25G A262686-500G A262686-1000G
76722-838EA 7 USD
76722-838 76722-840 76722-842 77601-124
Stearamide 98%
Stearamide

Precaución: for Research Use Only

Formula: C₁₈H₃₇NO
MDL Number: MFCD00008038
CAS Number: 124-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR