Sodium 5-(difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide 98%
Supplier: Ambeed
|
Warning
|
A261610-100MG
A261610-1G
A261610-1MG
A261610-250MG
A261610-5G
76722-810EA
33.95
USD
76722-810
76722-812
76722-814
76722-816
76722-818
Sodium 5-(difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide 98%
Sodium 5-(difluoromethoxy)-2-(((3,4-dimethoxypyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide
Precaución: for Research Use Only
Formula:
C₁₆H₁₄F₂N₃NaO₄S MW: 405.35 g/mol |
MDL Number:
MFCD01658543 CAS Number: 138786-67-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...