2,4,6-Trichloro-5-ethylpyrimidine 95%

Supplier: Ambeed
Warning

A260719-1G A260719-250MG
76722-796EA 442 USD
76722-796 76722-798
2,4,6-Trichloro-5-ethylpyrimidine 95%
2,4,6-Trichloro-5-ethylpyrimidine

Precaución: for Research Use Only

Formula: C₆H₅Cl₃N₂
MDL Number: MFCD11112097
CAS Number: 1780-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR