SA4503 dihydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Cutamesine dihydrochloride, AGY94806 dihydrochloride, 1-[2-(3,4-dimethoxyphenyl)ethyl]-4-(3-phenylpropyl)piperazine dihydrochloride, 1-(3,4-Dimethoxyphenethyl)-4-(3-phenylpropyl)piperazine 2HCl

A260566-10MG A260566-1MG A260566-50MG A260566-5MG
76722-780EA 87 USD
76722-780 76722-782 76722-784 76722-786
SA4503 dihydrochloride 98%
SA4503 dihydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₃₄Cl₂N₂O₂
MW: 441.43 g/mol
MDL Number: MFCD00939308
CAS Number: 165377-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR