Octyltriethoxysilane 97%

Supplier: Ambeed
Warning

Synonyms: Triethoxy(octyl)silane, Triethoxyoctylsilane

A260155-100G A260155-25G A260155-500G A260155-1000G
76722-774EA 12.74 USD
76722-774 76722-776 76722-778 77593-622
Octyltriethoxysilane 97%
Octyltriethoxysilane

Precaución: for Research Use Only

Formula: C₁₄H₃₂O₃Si
MW: 276.49 g/mol
MDL Number: MFCD00039883
CAS Number: 2943-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR