N,N-Dimethyl-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(Dimethylamino)-3-phenylpropanoic acid, L-Phenylalanine, N,N-dimethyl-, Alanine, N,N-dimethyl-3-phenyl-, L-

A259876-1G A259876-250MG A259876-25G A259876-5G
76722-766EA 22.28 USD
76722-766 76722-768 76722-770 76722-772
N,N-Dimethyl-L-phenylalanine 95%
N,N-Dimethyl-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₂
MW: 193.246 g/mol
MDL Number: MFCD00082539
CAS Number: 17469-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR