2-Amino-4'-bromobenzophenone 98%

Supplier: Ambeed
Warning

A259558-10G A259558-1G A259558-25G A259558-5G
76722-752EA 64 USD
76722-752 76722-754 76722-756 76722-758
2-Amino-4'-bromobenzophenone 98%
2-Amino-4'-bromobenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrNO
MDL Number: MFCD02181034
CAS Number: 1140-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR