Methyl oxalyl chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate
A259069-10G
A259069-25G
A259069-100G
76722-748EA
8.49
USD
76722-748
76722-750
77668-116
Methyl oxalyl chloride 98%
Methyl oxalyl chloride
Precaución: for Research Use Only
Formula:
C₃H₃ClO₃ MW: 122.51 g/mol |
MDL Number:
MFCD00000705 CAS Number: 5781-53-3 UN: 2920 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...