4-Chloro-5'-fluoro-2'-hydroxybenzophenone 97%

Supplier: Ambeed
Warning

A258299-250MG A258299-5G
76722-728EA 17 USD
76722-728 77872-272
4-Chloro-5'-fluoro-2'-hydroxybenzophenone 97%
4-Chloro-5'-fluoro-2'-hydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₈ClFO₂
MDL Number: MFCD00040948
CAS Number: 62433-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR