5-Acetamido-2-hydroxybenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-(Acetylamino)-2-hydroxybenzoic acid, N-Acetyl mesalazine, 5-Acetamidosalicylic acid

A258226-100MG A258226-250MG A258226-1G A258226-5G
76722-724EA 47 USD
76722-724 76722-726 77668-108 77879-484
5-Acetamido-2-hydroxybenzoic acid 98%
5-Acetamido-2-hydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉NO₄
MDL Number: MFCD00463101
CAS Number: 51-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR