Rhodium(III) iodide 98%

Supplier: Ambeed
Warning

A257936-100MG A257936-250MG
76722-710EA 33 USD
76722-710 76722-712
Rhodium(III) iodide 98%
Rhodium(III) iodide

Precaución: for Research Use Only

Formula: I₃Rh
MW: 483.62
Boiling Pt: 100
Density: 6.4
MDL Number: MFCD00016311
CAS Number: 15492-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR