4-Amino-N-phenylbenzamide 97%

Supplier: Ambeed
Warning

A256837-100MG A256837-1G A256837-250MG
76722-692EA 9.55 USD
76722-692 76722-694 76722-696
4-Amino-N-phenylbenzamide 97%
4-Amino-N-phenylbenzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₂N₂O
MW: 212.2472 g/mol
MDL Number: MFCD00017107
CAS Number: 782-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR