Methyl-5-methoxy-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-methoxy-2-nitrobenzoate, 5-Methoxy-2-nitrobenzoic acid methyl ester

A255925-10G A255925-1G A255925-5G
76722-664EA 50.93 USD
76722-664 76722-666 77668-072
Methyl-5-methoxy-2-nitrobenzoate 95%
Methyl-5-methoxy-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD00662902
CAS Number: 2327-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR