Methyl-5-iodsalicylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-hydroxy-5-iodobenzoate

A255677-100G A255677-1G A255677-5G
76722-658EA 259.93 USD
76722-658 76722-660 76722-662
Methyl-5-iodsalicylate 98%
Methyl 5-iodsalicylate

Precaución: for Research Use Only

Formula: C₈H₇IO₃
MW: 278.05 g/mol
MDL Number: MFCD00016462
CAS Number: 4068-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR