Oseltamivir phosphate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3R,4R,5S)-Ethyl 4-acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-enecarboxylate phosphate
A255018-100MG
A255018-1G
A255018-1MG
A255018-25MG
A255018-5G
A255018-250MG
A255018-100G
76722-646EA
9
USD
76722-646
76722-648
76722-650
76722-652
77601-052
77668-070
77879-468
Oseltamivir phosphate 98%
Oseltamivir phosphate
Precaución: for Research Use Only
Formula:
C₁₆H₂₈N₂O₄·H₃PO₄ MW: 410.40 g/mol |
MDL Number:
MFCD08059548 CAS Number: 204255-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...