Piperlongumine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one
A250732-100MG
A250732-10MG
A250732-1G
A250732-1MG
A250732-25MG
A250732-50MG
A250732-5MG
A250732-250MG
A250732-5G
76722-614EA
90.18
USD
76722-614
76722-616
76722-618
76722-620
76722-622
76722-624
76722-626
77668-054
77668-056
Piperlongumine 98%
Piperlongumine
Precaución: for Research Use Only
Formula:
C₁₇H₁₉NO₅ MW: 317.341 g/mol |
MDL Number:
MFCD00075706 CAS Number: 20069-09-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...