Methyl-3,4,5-trihydroxybenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl gallate, Gallic acid methyl ester, Methyl 3,4,5-trihydroxybenzoate

A249559-100G A249559-10G A249559-500G A249559-25G A249559-1000G
76722-594EA 10.61 USD
76722-594 76722-596 76722-598 77304-490 77668-044
Methyl-3,4,5-trihydroxybenzoate 98%
Methyl-3,4,5-trihydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₈O₅
MW: 184.15 g/mol
MDL Number: MFCD00002194
CAS Number: 99-24-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR