Methyl-α-D-glucopyranoside 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
A248611-1000G
A248611-100G
A248611-25G
A248611-500G
76722-582EA
56.23
USD
76722-582
76722-584
76722-586
76722-588
Methyl-α-D-glucopyranoside 98%
Methyl-α-D-glucopyranoside
Precaución: for Research Use Only
Formula:
C₇H₁₄O₆ MW: 194.18 g/mol |
MDL Number:
MFCD00064086 CAS Number: 97-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...