O-Phenyl chlorothioformate 95%

Supplier: Ambeed
Danger

Synonyms: Phenyl chlorothionocarbonate, Phenyl chlorothionoformate, O-Phenyl carbonochloridothioate

A248164-100G A248164-10G A248164-1G A248164-25G A248164-500G A248164-5G
76722-570EA 143 USD
76722-570 76722-572 76722-574 76722-576 76722-578 76722-580
O-Phenyl chlorothioformate 95%
O-Phenyl chlorothioformate

Precaución: for Research Use Only

Formula: C₇H₅ClOS
MDL Number: MFCD00004920
CAS Number: 1005-56-7
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR