Octadecyltriethoxysilane 85%

Supplier: Ambeed
Warning

Synonyms: Triethoxy(octadecyl)silane

A246942-25G A246942-5G A246942-100G
76722-548EA 37 USD
76722-548 76722-550 77879-448
Octadecyltriethoxysilane 85%
Octadecyltriethoxysilane

Precaución: for Research Use Only

Formula: C₂₄H₅₂O₃Si
MW: 416.76 g/mol
MDL Number: MFCD00039875
CAS Number: 7399-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR