5,7-Dihydroxy-6-methoxy-2-phenyl-4H-chromen-4-one 98%

Supplier: Ambeed
Warning

Synonyms: Oroxylin A

A246075-5MG A246075-100MG
76722-540EA 6.37 USD
76722-540 77668-006
5,7-Dihydroxy-6-methoxy-2-phenyl-4H-chromen-4-one 98%
5,7-Dihydroxy-6-methoxy-2-phenyl-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₆H₁₂O₅
MW: 284.26 g/mol
MDL Number: MFCD02259441
CAS Number: 480-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR