N-(tert-Butyl)hydroxylamine hydrochloride 95%

Supplier: Ambeed
Warning

A245257-100G A245257-10G A245257-1G A245257-250MG A245257-25G A245257-5G
76722-518EA 207 USD
76722-518 76722-520 76722-522 76722-524 76722-526 76722-528
N-(tert-Butyl)hydroxylamine hydrochloride 95%
N-(tert-Butyl)hydroxylamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₁NO
MDL Number: MFCD00012598
CAS Number: 57497-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR