Methyl phenoxyacetate 98%

Supplier: Ambeed
Warning

A244754-1000G A244754-100G A244754-10G A244754-25G
76722-504EA 180 USD
76722-504 76722-506 76722-508 76722-510
Methyl phenoxyacetate 98%
Methyl phenoxyacetate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00010227
CAS Number: 2065-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR