L(+)-Potassium sodium tartrate tetrahydrate 95%

Supplier: Ambeed
Warning

Synonyms: Potassium sodium tartrate tetrahydrate, Potassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate, Potassium sodium L(+)-tartrate tetrahydrate, Sodium potassium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrate, (R,R)-(+)-Potassium sodium tartrate tetrahydrate, Rochelle salt, L(+)-Tartaric acid potassium sodium salt tetrahydrate, (+)-Potassium sodium L-tartrate tetrahydrate

A244468-100G A244468-500G A244468-1000G
76722-500EA 6 USD
76722-500 76722-502 77550-566
L(+)-Potassium sodium tartrate tetrahydrate 95%
L(+)-Potassium sodium tartrate tetrahydrate

Precaución: for Research Use Only

Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
MDL Number: MFCD00150989
CAS Number: 6381-59-5
Merck Index: 13,07755

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR