Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide 99%

Supplier: Ambeed
Warning

Synonyms: BAPO, Bis-trimethylbenzoyl phenylphosphine oxide, Bisacylphosphine oxides, [phenyl-(2,4,6-trimethylbenzoyl)phosphoryl]-(2,4,6-trimethylphenyl)methanone, (Phenylphosphoryl)bis[(2,4,6-trimethylphenyl)methanone]

A244281-100G A244281-25G A244281-500G A244281-1000G
76722-492EA 16 USD
76722-492 76722-494 76722-496 76722-498 77879-431
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide 99%
Phenylbis(2,4,6-trimethylbenzoyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₆H₂₇O₃P
MW: 418.47 g/mol
MDL Number: MFCD01863675
CAS Number: 162881-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR