Nimustine hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: Nimustine HCl, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-1-(2-chloroethyl)-1-nitrosourea hydrochloride, ACNU, 1-[(4-Amino-2-methyl-5-pyrimidinyl)methyl]-3-(2-chloroethyl)-3-nitrosourea Hydrochloride

A243509-100MG A243509-50MG
76722-474EA 32 USD
76722-474 76722-476 76722-478 76722-480 76722-482 76722-484
Nimustine hydrochloride 97%
Nimustine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃ClN₆O₂
MW: 309.15 g/mol
MDL Number: MFCD01676942
CAS Number: 55661-38-6
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR