2-Amino-4-(1-naphthyl)thiazole 98%

Supplier: Ambeed
Warning

Synonyms: 4-(Naphthalen-1-yl)thiazol-2-amine

A243325-1G A243325-250MG A243325-25G A243325-5G
76722-466EA 35.01 USD
76722-466 76722-468 76722-470 76722-472
2-Amino-4-(1-naphthyl)thiazole 98%
2-Amino-4-(1-naphthyl)thiazole

Precaución: for Research Use Only

Formula: C₁₃H₁₀N₂S
MW: 226.30 g/mol
MDL Number: MFCD00236016
CAS Number: 56503-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR