7-Chloropyrido[3,4-b]pyrazine 95%

Supplier: Ambeed
Warning

A241232-100MG A241232-1G A241232-250MG A241232-5G
76722-450EA 60 USD
76722-450 76722-452 76722-454 76722-456
7-Chloropyrido[3,4-b]pyrazine 95%
7-Chloropyrido[3,4-b]pyrazine

Precaución: for Research Use Only

Formula: C₇H₄ClN₃
MDL Number: MFCD11044722
CAS Number: 93049-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR