4-Methylmorpholine hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A241185-25G
A241185-100G
A241185-1G
A241185-5G
76722-448EA
189
USD
76722-448
77600-944
77879-424
77879-425
4-Methylmorpholine hydrochloride 98%
4-Methylmorpholine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂ClNO |
MDL Number:
MFCD00039042 CAS Number: 3651-67-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...