Sodium dichloroacetate 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 2,2-dichloroacetate

A239769-100G A239769-100MG A239769-10G A239769-1G A239769-25G A239769-5G
76722-418EA 206.88 USD
76722-418 76722-420 76722-422 76722-424 76722-426 76722-428
Sodium dichloroacetate 97%
Sodium dichloroacetate

Precaución: for Research Use Only

Formula: C₂HCl₂NaO₂
MW: 150.92 g/mol
MDL Number: MFCD00070489
CAS Number: 2156-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR