Methyl quinoline-3-carboxylate 97%

Supplier: Ambeed
Warning

A238536-250MG A238536-5G A238536-1G A238536-25G
76722-380EA 18 USD
76722-380 76722-382 77568-306 77879-415
Methyl quinoline-3-carboxylate 97%
Methyl quinoline-3-carboxylate

Precaución: For research use only.

Formula: C₁₁H₉NO₂
MDL Number: MFCD03550859
CAS Number: 53951-84-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR