Camostat methanesulfate 95%

Supplier: Ambeed
Danger

Synonyms: Camostat Mesylate, Camostat Methanesulphonate, 4-(2-(2-(Dimethylamino)-2-oxoethoxy)-2-oxoethyl)phenyl 4-guanidinobenzoate methanesulphonate

A238171-100MG A238171-10MG A238171-1G A238171-250MG A238171-50MG A238171-5G A238171-1MG
76722-354EA 30.77 USD
76722-354 76722-356 76722-358 76722-360 76722-362 76722-364 77600-910
Camostat methanesulfate 95%
Camostat methanesulphate

Precaución: for Research Use Only

Formula: C₂₀H₂₂N₄O₅·CH₃SO₃H
MW: 494.52 g/mol
MDL Number: MFCD00941410
CAS Number: 59721-29-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR