Potassium hexacyanoferrate(II) trihydrate 99%

Supplier: Ambeed
Warning

A235710-1000G A235710-100G A235710-25G A235710-500G
76722-326EA 22.28 USD
76722-326 76722-328 76722-330 76722-332
Potassium hexacyanoferrate(II) trihydrate 99%
Potassium hexacyanoferrate(II) trihydrate

Precaución: for Research Use Only

Formula: K₄Fe(CN)₆·3H₂O
MDL Number: MFCD00167023
CAS Number: 14459-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR