N-Benzoyl-DL-tyrosyl-N',N'-dipropylamide 97%

Supplier: Ambeed
Warning

Synonyms: N-(1-(Dipropylamino)-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)benzamide

A233298-10G A233298-1G A233298-250MG A233298-25G A233298-5G
76722-306EA 39 USD
76722-306 76722-308 76722-310 76722-312 76722-314
N-Benzoyl-DL-tyrosyl-N',N'-dipropylamide 97%
N-Benzoyl-DL-tyrosyl-N',N'-dipropylamide

Precaución: for Research Use Only

Formula: C₂₂H₂₈N₂O₃
MDL Number: MFCD08063358
CAS Number: 57227-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR