Magnesium stearate Mg(4-5%)

Supplier: Ambeed

A232605-100G A232605-25G A232605-500G
76722-300EA 6.37 USD
76722-300 76722-302 76722-304
Magnesium stearate Mg(4-5%)
Magnesium stearate
Formula: C₃₆H₇₀MgO₄
MDL Number: MFCD00036391
CAS Number: 557-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR