Montelukast sodium 98%

Supplier: Ambeed
Warning

A232574-100MG A232574-10MG A232574-1G A232574-250MG A232574-25G A232574-50MG A232574-1MG A232574-5G
76722-288EA 15 USD
76722-288 76722-290 76722-292 76722-294 76722-296 76722-298 77667-916 77667-918
Montelukast sodium 98%
Montelukast sodium

Precaución: for Research Use Only

Formula: C₃₅H₃₆ClNO₃S
MDL Number: MFCD00931431
CAS Number: 151767-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR