9-Phenanthrenol 95%

Supplier: Ambeed
Warning

Synonyms: 9-Hydroxyphenanthrene, Phenanthren-9-ol, 9-Phenanthrol

A232543-1G A232543-1MG A232543-250MG A232543-5G A232543-25G A232543-100MG
76722-280EA 169.75 USD
76722-280 76722-282 76722-284 76722-286 77600-868 77667-914
9-Phenanthrenol 95%
9-Phenanthrenol

Precaución: for Research Use Only

Formula: C₁₄H₁₀O
MW: 194.23 g/mol
MDL Number: MFCD00082298
CAS Number: 484-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR