3-Carboxy-5-nitrophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 3-Carboxy-5-nitrobenzeneboronic acid pinacol ester, 3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-nitrophenylboronic acid, pinacol ester

A232486-100MG A232486-1G A232486-250MG A232486-5G
76722-272EA 9 USD
76722-272 76722-274 76722-276 76722-278
3-Carboxy-5-nitrophenylboronic acid pinacol ester 97%
3-Carboxy-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₆
MDL Number: MFCD03411932
CAS Number: 377780-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR