Moexipril hydrochloride 98%

Supplier: Ambeed
Danger

Synonyms: (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-3-isoquinolinecarboxylic acid hydrochloride

A232444-100MG A232444-1G A232444-250MG
76722-266EA 113.52 USD
76722-266 76722-268 76722-270
Moexipril hydrochloride 98%
Moexipril hydrochloride

Precaución: for Research Use Only

Formula: C₂₇H₃₅N₂O₇Cl
MW: 535.03 g/mol
MDL Number: MFCD00940481
CAS Number: 82586-52-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR