2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98%

Supplier: Ambeed
Warning

A232232-100MG A232232-1G A232232-250MG
76722-260EA 57 USD
76722-260 77879-395 77879-396
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide 98%
2-Methyl-6-oxo-1,6-dihydro-3,4'-bipyridine-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₂H₁₁N₃O₂
MDL Number: MFCD09800601
CAS Number: 80047-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR