N-Me-Thr(tBu)-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Me-Thr(tBu)-OH

A231922-100MG A231922-1G A231922-250MG
76722-250EA 70.02 USD
76722-250 76722-252 76722-254
N-Me-Thr(tBu)-OH 95%
N-Me-Thr(tBu)-OH

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.255 g/mol
MDL Number: MFCD02094573
CAS Number: 42417-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR