Ethyl carbamimidate hydrochloride 97%

Supplier: Ambeed
Warning

A231774-10G A231774-1G A231774-25G A231774-5G
76722-238EA 59.42 USD
76722-238 76722-240 76722-242 76722-244
Ethyl carbamimidate hydrochloride 97%
Ethyl carbamimidate hydrochloride

Precaución: for Research Use Only

Formula: C₃H₉ClN₂O
MW: 124.5694 g/mol
MDL Number: MFCD07438010
CAS Number: 31407-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR