Ethyl 3-methylbenzofuran-2-carboxylate 97%

Supplier: Ambeed
Warning

A230576-1G A230576-25G A230576-5G
76722-226EA 10.61 USD
76722-226 76722-228 76722-230
Ethyl 3-methylbenzofuran-2-carboxylate 97%
Ethyl 3-methylbenzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₂O₃
MW: 204.22 g/mol
MDL Number: MFCD00846334
CAS Number: 22367-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR