(R)-Glycidyl trityl ether 98%

Supplier: Ambeed
Warning

A229573-1G A229573-25G A229573-5G
76722-214EA 12 USD
76722-214 76722-216 76722-218
(R)-Glycidyl trityl ether 98%
(R)-Glycidyl trityl ether

Precaución: for Research Use Only

Formula: C₂₂H₂₀O₂
MDL Number: MFCD00273368
CAS Number: 65291-30-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR