N,N'-Bis-Z-1-guanylpyrazole 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-Bis-Z-1-Guanylpyrazole

A229496-100G A229496-1G A229496-250MG A229496-25G A229496-5G
76722-204EA 402.09 USD
76722-204 76722-206 76722-208 76722-210 76722-212
N,N'-Bis-Z-1-guanylpyrazole 98%
N,N'-Bis-Z-1-guanylpyrazole

Precaución: for Research Use Only

Formula: C₂₀H₁₈N₄O₄
MW: 378.38 g/mol
MDL Number: MFCD02683516
CAS Number: 152120-55-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR