Nelfinavir mesylate 98%

Supplier: Ambeed
Warning

Synonyms: (3S,4aS,8aS)-N-(tert-Butyl)-2-((2R,3R)-2-hydroxy-3-(3-hydroxy-2-methylbenzamido)-4-(phenylthio)butyl)decahydroisoquinoline-3-carboxamide methanesulphonate, AG 1341

A228436-10MG A228436-1MG A228436-5MG
76722-190EA 110.34 USD
76722-190 76722-192 76722-194
Nelfinavir mesylate 98%
Nelfinavir mesylate

Precaución: for Research Use Only

Formula: C₃₂H₄₅N₃O₄S·CH₄O₃S
MW: 663.9 g/mol
MDL Number: MFCD00931436
CAS Number: 159989-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR